Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nr4a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010444.2 | NP_034574.1 | 5 | 1339 | ||
GenBank mRNA | AK004709.1 | - | 5 | 1333 | ||
AK157203.1 | - | 5 | 1339 | |||
BC004770.1 | - | 5 | 1319 | 3156434 | ||
CT010287.1 | - | 4 | 1211 | |||
J04113.1 | - | 5 | 1322 | |||
X60132.1 | - | 5 | 1289 |