Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hoxa1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010449.4 | NP_034579.3 | 1 | 580 | ||
GenBank mRNA | AK083575.1 | - | 1 | 581 | ||
BC138097.1 | - | 1 | 514 | 8861116 | ||
BC138098.1 | - | 1 | 517 | 8861117 | ||
M22115.1 | - | 1 | 549 |