Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eif2ak1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013557.2 | NP_038585.2 | 1322 | |||
GenBank mRNA | AK156492.1 | - | 1281 | |||
AK173160.1 | - | 1272 | ||||
AY033898.1 | - | 1307 | ||||
BC028923.1 | - | 1294 | 4164417 | |||
BC111035.1 | - | 1273 | 5357121 |