Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prmt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF169620.1 | - | 4 | 422 | ||
AK077456.1 | - | 4 | 479 | |||
AK140200.1 | - | 4 | 506 | |||
AK144412.1 | - | 4 | 477 | |||
AK145829.1 | - | 4 | 479 | |||
AK146754.1 | - | 4 | 478 | |||
AK146966.1 | - | 4 | 486 | |||
AK159186.1 | - | 4 | 485 | |||
BC088997.1 | - | 4 | 436 | |||
BC122563.1 | - | 3 | 410 | 40107557 | ||
BC125275.1 | - | 3 | 410 | 40107558 | ||
BY727303.1 | - | 4 | 505 |