Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prmt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF169620.1 | - | 5 | 502 | ||
AK077456.1 | - | 5 | 559 | |||
AK140200.1 | - | 5 | 586 | |||
AK144412.1 | - | 5 | 557 | |||
AK145829.1 | - | 5 | 559 | |||
AK146754.1 | - | 5 | 558 | |||
AK146966.1 | - | 5 | 566 | |||
AK159186.1 | - | 5 | 565 | |||
BC088997.1 | - | 5 | 516 | |||
BC122563.1 | - | 4 | 490 | 40107557 | ||
BC125275.1 | - | 4 | 490 | 40107558 | ||
BY727303.1 | - | 5 | 585 |