Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prmt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF169620.1 | - | 3 | 163 | ||
AK077456.1 | - | 3 | 220 | |||
AK140200.1 | - | 3 | 247 | |||
AK144412.1 | - | 3 | 218 | |||
AK145829.1 | - | 3 | 220 | |||
AK146754.1 | - | 3 | 219 | |||
AK146966.1 | - | 3 | 227 | |||
AK159186.1 | - | 3 | 226 | |||
BC088997.1 | - | 3 | 177 | |||
BC122563.1 | - | 2 | 151 | 40107557 | ||
BC125275.1 | - | 2 | 151 | 40107558 | ||
BY288156.1 | - | 3 | 255 | |||
BY727303.1 | - | 3 | 246 |