Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hs3st1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010474.2 | NP_034604.1 | 2 | 722 | ||
GenBank mRNA | AF019385.1 | - | 723 | |||
AK087753.1 | - | 2 | 701 | |||
AK132292.1 | - | 2 | 733 | |||
BC009133.1 | - | 2 | 684 | 3155049 |