Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hsd11b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001044751.1 | NP_001038216.1 | 4 | 470 | ||
NM_008288.2 | NP_032314.2 | 5 | 496 | |||
GenBank mRNA | AI099306.1 | - | 4 | 473 | ||
AK149518.1 | - | 4 | 468 | |||
AK165212.1 | - | 4 | 468 | |||
AK167419.1 | - | 3 | 420 | |||
BC132364.1 | - | 5 | 436 | 40130641 | ||
BC138780.1 | - | 5 | 436 | 8861802 | ||
BC145415.1 | - | 5 | 436 | 9053973 | ||
DQ089001.1 | - | 4 | 372 | |||
S75207.1 | - | 4 | 417 | |||
X83202.1 | - | 5 | 499 |