Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hsd11b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001044751.1 | NP_001038216.1 | 6 | 892 | ||
NM_008288.2 | NP_032314.2 | 7 | 918 | |||
GenBank mRNA | AI132676.1 | - | 1 | 460 | ||
AK149518.1 | - | 6 | 890 | |||
AK165212.1 | - | 6 | 890 | |||
AK167419.1 | - | 5 | 842 | |||
BC132364.1 | - | 7 | 858 | 40130641 | ||
BC138780.1 | - | 7 | 858 | 8861802 | ||
BC145415.1 | - | 7 | 858 | 9053973 | ||
DQ089001.1 | - | 6 | 794 | |||
S75207.1 | - | 6 | 839 | |||
X83202.1 | - | 7 | 921 |