Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Htr3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099644.1 | NP_001093114.1 | 3, 4 | 417 | ||
NM_013561.2 | NP_038589.2 | 3, 4 | 417 | |||
GenBank mRNA | AF513221.1 | - | 3, 4 | 294 | ||
AY605711.1 | - | 3, 4 | 283 | |||
BC144799.1 | - | 3, 4 | 339 | 9053347 | ||
BC145743.1 | - | 3, 4 | 339 | 40131046 | ||
BY274793.1 | - | 3, 4 | 418 | |||
M74425.1 | - | 3, 4 | 340 | |||
X72395.1 | - | 3, 4 | 301 |