Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Htr4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008313.4 | NP_032339.2 | 5 | 598 | ||
GenBank mRNA | AJ011369.1 | - | 4 | 388 | ||
AK141587.1 | - | 5 | 542 | |||
AK141625.1 | - | 5 | 599 | |||
BC132189.1 | - | 4 | 427 | |||
BC132191.1 | - | 4 | 427 | |||
BC145159.1 | - | 4 | 427 | |||
BC145161.1 | - | 4 | 427 | |||
JN418927.1 | - | 5 | 529 | |||
JN418928.1 | - | 5 | 515 | |||
JN418929.1 | - | 4 | 388 | |||
JN418930.1 | - | 4 | 404 | |||
Y09585.1 | - | 4 | 388 | |||
Y09587.1 | - | 4 | 388 | |||
Y09588.1 | - | 4 | 388 |
Set Membership: |
G-protein coupled receptor |