Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Htr4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008313.4 | NP_032339.2 | 5 | 659 | ||
GenBank mRNA | AJ011369.1 | - | 4 | 449 | ||
AK141587.1 | - | 5 | 603 | |||
AK141625.1 | - | 5 | 660 | |||
BC132189.1 | - | 4 | 488 | |||
BC132191.1 | - | 4 | 488 | |||
BC145159.1 | - | 4 | 488 | |||
BC145161.1 | - | 4 | 488 | |||
JN418927.1 | - | 5 | 590 | |||
JN418928.1 | - | 5 | 576 | |||
JN418929.1 | - | 4 | 449 | |||
JN418930.1 | - | 4 | 465 | |||
Y09585.1 | - | 4 | 449 | |||
Y09587.1 | - | 4 | 449 | |||
Y09588.1 | - | 4 | 449 |
Set Membership: |
G-protein coupled receptor |