Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Elavl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177883.1 | NP_001171354.1 | 2 | 638 | ||
NM_010486.2 | NP_034616.1 | 2 | 233 | |||
NM_207685.1 | NP_997568.1 | 3 | 345 | |||
NM_207686.1 | NP_997569.1 | 2 | 233 | |||
GenBank mRNA | AK141919.1 | - | 2 | 514 | ||
AK149201.1 | - | 2 | 450 | |||
AY035378.1 | - | 1 | 82 | |||
AY035379.1 | - | 1 | 82 | |||
AY035380.1 | - | 1 | 82 | |||
BC046598.1 | - | 2 | 157 | 6312245 | ||
BC049125.1 | - | 3 | 345 | 6821133 | ||
BC058393.1 | - | 3 | 345 | 6821133 | ||
U29088.1 | - | 2 | 232 |