Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ifnar1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010508.2 | NP_034638.2 | 8, 9 | 1254 | ||
GenBank mRNA | AK132431.1 | - | 8, 9 | 1186 | ||
AK136950.1 | - | 8, 9 | 1230 | |||
AK141630.1 | - | 8, 9 | 1190 | |||
AK148246.1 | - | 8, 9 | 1196 | |||
AK157555.1 | - | 9, 10 | 1282 | |||
BC043935.1 | - | 8, 9 | 1205 | 4507009 | ||
BC052217.1 | - | 8, 9 | 1214 | 30008497 | ||
BC052429.1 | - | 8, 9 | 1216 | 6836300 | ||
M89641.1 | - | 8, 9 | 1219 |