Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ifrd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013562.2 | NP_038590.2 | 11 | 1446 | ||
GenBank mRNA | AK078888.1 | - | 1444 | |||
AK168682.1 | - | 11 | 1442 | |||
AK183550.1 | - | 3 | 197 | |||
AK213011.1 | - | 2 | 93 | |||
AK215747.1 | - | 1 | 16 | |||
BC037013.1 | - | 11 | 1421 | |||
BC043723.1 | - | 11 | 1413 | 5369100 | ||
V00756.1 | - | 1 | 60 | |||
X17400.1 | - | 11 | 1423 |