Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Igf2r | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010515.2 | NP_034645.2 | 4 | 643 | ||
GenBank mRNA | BB865604.1 | - | 4 | 593 | ||
BC150817.1 | - | 4 | 554 | 9087728 | ||
BC171973.1 | - | 4 | 554 | 9054259 | ||
U04710.1 | - | 4 | 599 |