Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il10ra | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008348.2 | NP_032374.1 | 6, 7 | 881 | ||
GenBank mRNA | AK137663.1 | - | 6, 7 | 882 | ||
AK149680.1 | - | 6, 7 | 876 | |||
AK149899.1 | - | 5, 6 | 764 | |||
AK150022.1 | - | 6, 7 | 878 | |||
AK150524.1 | - | 6, 7 | 885 | |||
AK153120.1 | - | 6, 7 | 885 | |||
AK159566.1 | - | 6, 7 | 878 | |||
L12120.1 | - | 6, 7 | 886 |