Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il12b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF128214.1 | - | 2, 3 | 115 | ||
AF128215.1 | - | 2, 3 | 115 | |||
AK162981.1 | - | 2, 3 | 139 | |||
BC103608.1 | - | 2, 3 | 121 | 40046200 | ||
BC103609.1 | - | 2, 3 | 121 | 40046202 | ||
BC103610.1 | - | 2, 3 | 121 | 40046204 | ||
BC103614.1 | - | 2, 3 | 121 | 40046206 | ||
BY154173.1 | - | 2, 3 | 139 | |||
M86671.1 | - | 2, 3 | 116 |