Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il15ra | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271497.1 | NP_001258426.1 | 7 | 794 | ||
NM_001271498.1 | NP_001258427.1 | 7 | 888 | |||
NM_001271499.1 | NP_001258428.1 | 7 | 758 | |||
NM_001271500.1 | NP_001258429.1 | 6 | 659 | |||
NM_001271501.1 | NP_001258430.1 | 5 | 566 | |||
NM_008358.2 | NP_032384.1 | 7 | 857 | |||
NM_133836.2 | NP_598597.1 | 7 | 910 | |||
GenBank mRNA | AK040940.1 | - | 7 | 740 | ||
AK080000.1 | - | 7 | 740 | |||
AY219715.1 | - | 7 | 659 | |||
AY219716.1 | - | 6 | 561 | |||
AY219717.1 | - | 5 | 468 | |||
AY221616.1 | - | 7 | 535 | |||
AY221617.1 | - | 6 | 453 | |||
AY221618.1 | - | 5 | 360 | |||
AY221619.1 | - | 7 | 627 | |||
BC022705.1 | - | 7 | 667 | |||
BC069881.1 | - | 6 | 618 | |||
BC095982.1 | - | 4 | 585 | |||
BC132231.1 | - | 6 | 457 | |||
BC132233.1 | - | 6 | 457 | |||
BC145348.1 | - | 7 | 528 |