Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il7r | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008372.4 | NP_032398.3 | 5 | 770 | ||
GenBank mRNA | AF078906.1 | - | 5 | 636 | ||
AK040364.1 | - | 5 | 753 | |||
AK042264.1 | - | 5 | 715 | |||
AK154215.1 | - | 4 | 584 | |||
AK154516.1 | - | 5 | 730 | |||
BC089571.1 | - | 5 | 722 | 6758487 | ||
M29697.1 | - | 5 | 683 |