Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il7r | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008372.4 | NP_032398.3 | 1 | 150 | ||
GenBank mRNA | AF078906.1 | - | 1 | 16 | ||
AK040364.1 | - | 1 | 133 | |||
AK040740.1 | - | 1 | 133 | |||
AK041838.1 | - | 1 | 41 | |||
AK042264.1 | - | 1 | 95 | |||
AK154215.1 | - | 1 | 103 | |||
AK154516.1 | - | 1 | 110 | |||
BC089571.1 | - | 1 | 102 | 6758487 | ||
BY776757.1 | - | 1 | 151 | |||
M29697.1 | - | 1 | 63 |