Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ilf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042707.2 | NP_001036172.1 | 3 | 457 | ||
NM_001042708.2 | NP_001036173.1 | 4 | 496 | |||
NM_001042709.2 | NP_001036174.1 | 3 | 457 | |||
NM_001277321.1 | NP_001264250.1 | 4 | 496 | |||
NM_001277322.1 | NP_001264251.1 | 5 | 424 | |||
NM_010561.3 | NP_034691.2 | 4 | 496 | |||
GenBank mRNA | AB221609.1 | - | 2 | 172 | ||
AF098967.1 | - | 5 | 373 | |||
AF497751.2 | - | 4 | 364 | |||
AF497752.2 | - | 3 | 325 | |||
AK048096.1 | - | 3 | 387 | |||
AK077560.1 | - | 4 | 422 | |||
AK128936.1 | - | 4 | 429 | |||
AK150276.1 | - | 4 | 445 | |||
BB851524.1 | - | 3 | 460 | |||
BC047272.1 | - | 4 | 416 | 6440312 | ||
BC060599.1 | - | 2 | 160 | |||
DQ104405.1 | - | 4 | 364 | |||
DQ104406.1 | - | 3 | 325 |