Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ilf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042707.2 | NP_001036172.1 | 5 | 775 | ||
NM_001042708.2 | NP_001036173.1 | 6 | 814 | |||
NM_001042709.2 | NP_001036174.1 | 5 | 775 | |||
NM_001277321.1 | NP_001264250.1 | 6 | 814 | |||
NM_001277322.1 | NP_001264251.1 | 7 | 742 | |||
NM_010561.3 | NP_034691.2 | 6 | 814 | |||
GenBank mRNA | AB221609.1 | - | 4 | 490 | ||
AF098967.1 | - | 7 | 691 | |||
AF497751.2 | - | 6 | 682 | |||
AF497752.2 | - | 5 | 643 | |||
AK048096.1 | - | 5 | 705 | |||
AK077560.1 | - | 6 | 740 | |||
AK128936.1 | - | 6 | 747 | |||
AK150276.1 | - | 6 | 763 | |||
AK206785.1 | - | 2 | 86 | |||
BC047272.1 | - | 6 | 734 | 6440312 | ||
BC060599.1 | - | 4 | 478 | |||
DQ104405.1 | - | 6 | 682 | |||
DQ104406.1 | - | 5 | 643 |