Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ilf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042707.2 | NP_001036172.1 | 13 | 1786 | ||
NM_001042708.2 | NP_001036173.1 | 14 | 1825 | |||
NM_001042709.2 | NP_001036174.1 | 13 | 1786 | |||
NM_001277321.1 | NP_001264250.1 | 14 | 1825 | |||
NM_001277322.1 | NP_001264251.1 | 15 | 1753 | |||
NM_010561.3 | NP_034691.2 | 14 | 1825 | |||
GenBank mRNA | AB221609.1 | - | 12 | 1501 | ||
AF098967.1 | - | 15 | 1702 | |||
AF497751.2 | - | 14 | 1693 | |||
AF497752.2 | - | 13 | 1654 | |||
AK048096.1 | - | 13 | 1716 | |||
AK128936.1 | - | 14 | 1758 | |||
BC047272.1 | - | 14 | 1745 | 6440312 | ||
BC060599.1 | - | 11 | 1391 | |||
DQ104405.1 | - | 14 | 1693 | |||
DQ104406.1 | - | 13 | 1654 |