Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Itga2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008396.2 | NP_032422.2 | 15 | 1971 | ||
GenBank mRNA | AK051050.1 | - | 15 | 1982 | ||
AK156604.1 | - | 15 | 2013 | |||
BC065139.1 | - | 15 | 1971 | 30619401 | ||
X75427.1 | - | 4 | 516 | |||
Z29987.1 | - | 15 | 1975 |