Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Itgam | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001082960.1 | NP_001076429.1 | 18 | 2298 | ||
NM_008401.2 | NP_032427.2 | 18 | 2295 | |||
GenBank mRNA | AK040921.1 | - | 8 | 1024 | ||
AK089521.1 | - | 18 | 2262 | |||
AK146569.1 | - | 10 | 1200 | |||
AK154224.1 | - | 18 | 2215 | |||
AK154474.1 | - | 18 | 2249 | |||
AK154798.1 | - | 18 | 2211 | |||
AK155006.1 | - | 18 | 2227 | |||
AK155520.1 | - | 18 | 2220 | |||
AK155671.1 | - | 18 | 2264 | |||
AK155717.1 | - | 18 | 2253 | |||
AK170324.1 | - | 18 | 2267 | |||
AK171068.1 | - | 15 | 1927 |