Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Itk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281965.1 | NP_001268894.1 | 6 | 751 | ||
NM_001281966.1 | NP_001268895.1 | 6 | 742 | |||
NM_001281967.1 | NP_001268896.1 | 6 | 733 | |||
NM_001281968.1 | NP_001268897.1 | 6 | 730 | |||
NM_010583.3 | NP_034713.2 | 6 | 733 | |||
GenBank mRNA | AK083380.1 | - | 6 | 704 | ||
AK153613.1 | - | 6 | 689 | |||
BC028478.1 | - | 6 | 675 | |||
BC128374.1 | - | 6 | 659 | 40111687 | ||
BC128375.1 | - | 6 | 656 | 40111688 | ||
D14042.1 | - | 6 | 673 | |||
L00619.1 | - | 6 | 738 | |||
L05631.1 | - | 6 | 699 | |||
L10628.1 | - | 6 | 701 |