Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Jak3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190830.1 | NP_001177759.1 | 15, 16 | 2248 | ||
NM_010589.6 | NP_034719.2 | 15, 16 | 2248 | |||
GenBank mRNA | AK038268.1 | - | 14, 15 | 2059 | ||
AK088365.1 | - | 15, 16 | 2097 | |||
AK142178.1 | - | 15, 16 | 2086 | |||
AK156646.1 | - | 15, 16 | 2070 | |||
AK164520.1 | - | 15, 16 | 2088 | |||
BC105577.1 | - | 14, 15 | 2051 | 3968386 | ||
BC131646.1 | - | 14, 15 | 2059 | 40110146 | ||
BC131647.1 | - | 14, 15 | 2059 | 40110148 | ||
L32955.1 | - | 14, 15 | 2047 | |||
L33768.1 | - | 12, 13 | 2689 | |||
L40172.1 | - | 15, 16 | 2109 |