Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcna6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013568.6 | NP_038596.1 | 1 | 1342 | ||
GenBank mRNA | BC048782.1 | - | 1 | 824 | 6466414 | |
BC054804.1 | - | 1 | 862 | 6838056 | ||
M96688.1 | - | 1 | 1507 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AK031890.1 | - | 1107 | Chr. - - [-] on Build |