Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcnh2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001294162.1 | NP_001281091.1 | 7 | 1975 | ||
NM_013569.2 | NP_038597.2 | 11 | 3003 | |||
GenBank mRNA | AF012868.1 | - | 11 | 3009 | ||
AF012869.1 | - | 7 | 1923 | |||
AF034762.1 | - | 7 | 1967 | |||
AK145127.1 | - | 1980 | ||||
AY374424.1 | - | 11 | 2688 | |||
BC051016.1 | - | 11 | 2834 | |||
BC051233.1 | - | 1 | 41 | |||
CK619061.1 | - | 2 | 230 |