Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcnq2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB000494.1 | - | 1 | 311 | ||
AB000495.1 | - | 1 | 311 | |||
AB000496.1 | - | 1 | 311 | |||
AB000497.1 | - | 1 | 311 | |||
AB000498.1 | - | 1 | 311 | |||
AB000499.1 | - | 1 | 311 | |||
AB000500.1 | - | 1 | 311 | |||
AB000501.1 | - | 1 | 311 | |||
AB000502.1 | - | 1 | 311 | |||
AB000503.1 | - | 1 | 311 | |||
AB000504.1 | - | 1 | 311 | |||
AF490773.1 | - | 1 | 225 | |||
AK139411.1 | - | 1 | 56 | |||
AK161558.1 | - | 1 | 377 | |||
BC145453.1 | - | 1 | 355 | 9054011 | ||
BY726495.1 | - | 1 | 360 | |||
CJ119072.1 | - | 1 | 423 | |||
CJ121810.1 | - | 1 | 424 |