Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Napsa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008437.1 | NP_032463.1 | 3 | 483 | ||
GenBank mRNA | AB038144.1 | - | 3 | 496 | ||
AK002520.1 | - | 3 | 330 | |||
AK007861.1 | - | 3 | 329 | |||
AK152663.1 | - | 3 | 321 | |||
AK170013.1 | - | 3 | 327 | |||
BC014813.1 | - | 3 | 316 | 4221844 | ||
D88899.1 | - | 3 | 483 |
Gene Symbol: Gm15396 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK131623.1 | - | 2 | 88 |