Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Napsa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008437.1 | NP_032463.1 | 8 | 1149 | ||
GenBank mRNA | AK002520.1 | - | 8 | 996 | ||
AK152663.1 | - | 8 | 987 | |||
AK170013.1 | - | 8 | 993 | |||
BC014813.1 | - | 8 | 982 | 4221844 | ||
D88899.1 | - | 8 | 1149 |