Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kif13a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010617.2 | NP_034747.2 | 3, 4 | 261 | ||
GenBank mRNA | AB037923.2 | - | 3, 4 | 262 | ||
AK077053.1 | - | 3, 4 | 282 |