Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kif16b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081133.2 | NP_001074602.1 | 15, 16 | 1702 | ||
GenBank mRNA | AB610968.1 | - | 15, 16 | 1757 | ||
AK033018.1 | - | 9, 10 | 1029 | |||
AK129401.1 | - | 2, 3 | 153 |