Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kif3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290805.1 | NP_001277734.1 | 6 | 790 | ||
NM_001290806.1 | NP_001277735.1 | 6 | 790 | |||
NM_008443.4 | NP_032469.2 | 6 | 790 | |||
GenBank mRNA | AK147078.1 | - | 6 | 740 | ||
AK161223.1 | - | 6 | 781 | |||
AK161920.1 | - | 6 | 764 | |||
AK169425.1 | - | 6 | 764 | |||
BC023936.1 | - | 6 | 740 | |||
BC044720.1 | - | 6 | 751 | |||
BC052707.1 | - | 6 | 756 | 5702415 | ||
D12645.1 | - | 6 | 766 |