Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kif3c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008445.2 | NP_032471.2 | 1 | 2079 | ||
GenBank mRNA | AF013116.1 | - | 1 | 1655 | ||
AK133649.1 | - | 1 | 2061 | |||
AK147572.1 | - | 1 | 1881 | |||
AK178532.1 | - | 1 | 255 | |||
AK220283.1 | - | 2 | 1804 | |||
BC052732.1 | - | 1 | 1236 | |||
BC053053.1 | - | 1 | 2021 | |||
BC062648.1 | - | 1 | 1754 | |||
BC127063.1 | - | 1 | 1291 | 40109965 |