Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kif3c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008445.2 | NP_032471.2 | 2, 3 | 2491 | ||
GenBank mRNA | AF013116.1 | - | 2, 3 | 2067 | ||
AK139894.1 | - | 2, 3 | 167 | |||
AK147572.1 | - | 2, 3 | 2293 | |||
AK220283.1 | - | 3, 4 | 2216 | |||
BC052732.1 | - | 2, 3 | 1648 | |||
BC053053.1 | - | 2, 3 | 2433 | |||
BC062648.1 | - | 2, 3 | 2166 | |||
BC127063.1 | - | 2, 3 | 1703 | 40109965 |