Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025358.2 | NP_001020529.2 | 2 | 242 | ||
NM_001025359.2 | NP_001020530.2 | 2 | 242 | |||
NM_001025360.2 | NP_001020531.2 | 2 | 261 | |||
NM_001025361.2 | NP_001020532.2 | 2 | 242 | |||
NM_001025362.2 | NP_001020533.2 | 2 | 242 | |||
NM_001025363.2 | NP_001020534.2 | 2 | 242 | |||
NM_001081959.1 | NP_001075428.1 | 2 | 261 | |||
NM_008450.2 | NP_032476.2 | 2 | 261 | |||
GenBank mRNA | AF055665.1 | - | 2 | 160 | ||
AK031309.1 | - | 2 | 233 | |||
AK184188.1 | - | 2 | 116 | |||
AK207464.1 | - | 2 | 116 | |||
AU035615.1 | - | 2 | 261 | |||
AY753300.1 | - | 1 | 66 | |||
BC055744.1 | - | 2 | 97 | 6415280 | ||
BY014721.1 | - | 2 | 242 | |||
CF615940.1 | - | 2 | 172 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AK144708.1 | - | 226 | Chr. - - [-] on Build |