Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Krt27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010666.1 | NP_034796.1 | 8 | 1331 | ||
GenBank mRNA | AB013607.1 | - | 8 | 1331 | ||
AK077384.1 | - | 8 | 1343 | |||
BC140996.1 | - | 8 | 1355 | 9055608 | ||
BC140997.1 | - | 8 | 1355 | 9055609 |