Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Krt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010668.2 | NP_034798.2 | 3, 4 | 1003 | ||
GenBank mRNA | AK132476.1 | - | 3, 4 | 1005 | ||
AK161078.1 | - | 3, 4 | 1002 | |||
AK161098.1 | - | 3, 4 | 1005 | |||
BC120485.1 | - | 3, 4 | 969 | 40129408 | ||
BC139485.1 | - | 3, 4 | 969 | 8860587 | ||
X74784.1 | - | 3, 4 | 969 |