Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arhgef1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130150.1 | NP_001123622.1 | 17, 18 | 1760 | ||
NM_001130151.1 | NP_001123623.1 | 16, 17 | 1751 | |||
NM_001130152.1 | NP_001123624.1 | 15, 16 | 1592 | |||
NM_001130153.1 | NP_001123625.1 | 15, 16 | 1583 | |||
NM_008488.2 | NP_032514.1 | 15, 16 | 1580 | |||
GenBank mRNA | AK012156.1 | - | 15, 16 | 1534 | ||
AK043027.1 | - | 16, 17 | 1716 | |||
AK051518.1 | - | 15, 16 | 1533 | |||
AK131181.1 | - | 15, 16 | 1480 | |||
AK157056.1 | - | 17, 18 | 1696 | |||
AK172354.1 | - | 17, 18 | 1696 | |||
BC012488.1 | - | 15, 16 | 1543 | 3154558 | ||
U58203.1 | - | 15, 16 | 1510 |