Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arhgef1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130150.1 | NP_001123622.1 | 21 | 2133 | ||
NM_001130151.1 | NP_001123623.1 | 20 | 2124 | |||
NM_001130152.1 | NP_001123624.1 | 19 | 1965 | |||
NM_001130153.1 | NP_001123625.1 | 19 | 1956 | |||
NM_008488.2 | NP_032514.1 | 19 | 1953 | |||
GenBank mRNA | AK012156.1 | - | 19 | 1907 | ||
AK043027.1 | - | 20 | 2089 | |||
AK051518.1 | - | 19 | 1906 | |||
AK131181.1 | - | 19 | 1853 | |||
AK157056.1 | - | 21 | 2069 | |||
AK172354.1 | - | 21 | 2069 | |||
AK201417.1 | - | 1 | 15 | |||
BC012488.1 | - | 19 | 1916 | 3154558 | ||
U58203.1 | - | 19 | 1883 |