Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lepr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122899.1 | NP_001116371.1 | 8 | 1809 | ||
NM_010704.2 | NP_034834.1 | 8 | 1809 | |||
NM_146146.2 | NP_666258.2 | 8 | 1809 | |||
GenBank mRNA | AK140785.1 | - | 8 | 1311 | ||
AK143989.1 | - | 8 | 1498 | |||
AK155891.1 | - | 8 | 1349 | |||
BC082551.1 | - | 8 | 1294 | 30621706 | ||
U42467.1 | - | 8 | 1317 | |||
U46135.1 | - | 8 | 1317 | |||
U49106.1 | - | 7 | 1257 | |||
U49109.1 | - | 7 | 1257 | |||
U49110.1 | - | 7 | 1257 | |||
U52915.1 | - | 8 | 1771 | |||
U58861.1 | - | 8 | 1339 | |||
U58862.1 | - | 8 | 1349 | |||
U58863.1 | - | 8 | 1339 | |||
Y10296.1 | - | 7 | 1265 | |||
Y10298.1 | - | 7 | 1265 |