Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lig3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291245.1 | NP_001278174.1 | 3 | 811 | ||
NM_001291246.1 | NP_001278175.1 | 3 | 811 | |||
NM_001291247.1 | NP_001278176.1 | 3 | 815 | |||
NM_010716.3 | NP_034846.2 | 3 | 815 | |||
GenBank mRNA | AK010719.1 | - | 3 | 726 | ||
AK031540.1 | - | 3 | 753 | |||
AK143531.1 | - | 3 | 681 | |||
AK150644.1 | - | 3 | 723 | |||
AK156018.1 | - | 3 | 733 | |||
AK167363.1 | - | 3 | 723 | |||
BC026143.1 | - | 3 | 786 | |||
BC049240.1 | - | 3 | 718 | 4504384 | ||
BC083500.1 | - | 3 | 714 | 3673103 | ||
U66057.1 | - | 3 | 740 | |||
U66058.1 | - | 3 | 740 |