Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Limk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK139231.1 | - | 2 | 2630 | ||
AK141813.1 | - | 9 | 1285 | |||
AK170166.1 | - | 8 | 1060 | |||
AK172083.1 | - | 8 | 1044 | |||
BC063768.1 | - | 9 | 1265 | |||
U14166.1 | - | 9 | 1308 | |||
U15159.1 | - | 9 | 1280 |