Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008512.2 | NP_032538.2 | 43, 44 | 7697 | ||
GenBank mRNA | AF367720.1 | - | 43, 44 | 7668 | ||
X67469.1 | - | 43, 44 | 7677 |