Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrp6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008514.4 | NP_032540.2 | 2 | 313 | ||
GenBank mRNA | AF074265.1 | - | 2 | 182 | ||
BC060704.1 | - | 2 | 313 | 6409058 |