Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lta4h | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008517.2 | NP_032543.2 | 12, 13 | 1325 | ||
GenBank mRNA | AK134931.1 | - | 12, 13 | 1311 | ||
AK219839.1 | - | 1, 2 | 96 | |||
BC021417.1 | - | 12, 13 | 1319 | 5004113 | ||
M63848.1 | - | 1291 |