Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stard4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133774.4 | NP_598535.1 | 6 | 740 | ||
GenBank mRNA | AF480297.1 | - | 6 | 595 | ||
AF480298.1 | - | 6 | 595 | |||
AK014587.1 | - | 1 | 535 | |||
AK042347.1 | - | 6 | 744 | |||
AK042691.1 | - | 6 | 700 | |||
AK054501.1 | - | 6 | 741 | |||
AK078858.1 | - | 6 | 744 | |||
AK085331.1 | - | 6 | 744 | |||
AK205539.1 | - | 3 | 378 | |||
BC005642.1 | - | 6 | 740 | 3709232 |